Discandiumtrioxid ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Synonyms: Scandium oxide, Scandium (III) oxide
11216.03 11216.06 11216.14
11216.03EA 1925 SEK
11216.03 11216.06 11216.14
Discandiumtrioxid ≥99.9% (REO, rare earth oxide basis), REacton®
Discandiumtrioxid
Purity: 99.9% CAS No.: 12060-08-1 Molecular Formula: Sc2O3 Formula Weight: 137.91 Physical Form: Powder Refractive Index: 1.964 Odour: Odourless Melting Point: 2485° Merck Reference: 14,8392 MDL No.: MFCD00011223
Formel: O₃Sc₂
MW: 137.91 g/mol
Smältpunkt: 2485 °C
Densitet: 3.86 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011223
CAS nummer: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,8392