Yttrium(III) fluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Yttrium trifluoride, Yttrium(III) fluoride
11178.14 11178.22 11178.36
11178.14EA 2850 SEK
11178.14 11178.22 11178.36
Yttrium(III) fluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium(III) fluoride
Purity: 99.99% CAS No.: 13709-49-4 Molecular Formula: YF3 Formula Weight: 145.90 Physical Form: Lump/powder Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049714
Formel: F₃Y
MW: 145.9 g/mol
Kokpunkt: 2227
Smältpunkt: 1387
Densitet: 4.01 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00049714
CAS nummer: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1 III