Kvicksilver(II)tiocyanat
Leverantör: Thermo Fisher Scientific
|
Fara
|
CAS No.: 592-85-8
Molecular Formula: C2HgN2S2
Formula Weight: 316.75
Physical Form: Powder
Density: 3.71
Sensitivity: Hygroscopic
Appearance: White to pale yellow
Odour: Odourless
Melting Point: ca 165° dec.
Beilstein Reference: 3561344
Merck Reference: 14,5890
UN No.: 1646
MDL No.: MFCD00011039
Formel:
C₂HgN₂S₂ Smältpunkt: ∼ 165 °C (dec.) Densitet: 3,71 Flampunkt: 120 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00011039 CAS nummer: 592-85-8 EINECS: 209-773-0 UN: 1646 ADR: 6.1,II Merck Index: 14,5890 |