Kvicksilver(II)tiocyanat 99+%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Mercury dithiocyanate, Mercuric thiocyanate, Mercury (II) rhodanide
196890500 196892500
ACRO196890500EA 1450 SEK
ACRO196890500 ACRO196892500
Kvicksilver(II)tiocyanat 99+%
Kvicksilver(II)tiocyanat
Formel: C₂HgN₂S₂
MW: 316.76 g/mol
Smältpunkt: 165 °C (dec.)
Flampunkt: 120
Storage Temperature: Omgivande
MDL Number: MFCD00011039
CAS nummer: 592-85-8
EINECS: 209-773-0
UN: 1646
ADR: 6.1 II
Merck Index: 13,05917
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Titration Complexometric ≥99.0 %
Iron (Fe) ≤20 ppm