Zinksulfid 99.99% < 10 µm
Leverantör: Thermo Scientific
Synonyms: Zinc (II) sulphide
223651000 223650250 223655000
ACRO223651000EA 1875 SEK
ACRO223651000 ACRO223650250 ACRO223655000
Zinksulfid 99.99% < 10 µm
Zinksulfid
Formel: SZn
MW: 97,46 g/mol
Smältpunkt: 1064 °C
Densitet: 4,1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011301
CAS nummer: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294
Order Now
Specifikation testresultat
Appearance White to light yellow Powder
Total trace metal impurities ≤0.01 %