Kvicksilver(II)oxid, gul 99+%
Leverantör: Thermo Fisher Scientific
Fara
205731000 205735000
ACRO205731000EA 1225 SEK
ACRO205731000 ACRO205735000
Kvicksilver(II)oxid, gul 99+%
Kvicksilver(II)oxid
Formel: HgO
Smältpunkt: 500 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011045
CAS nummer: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II
Merck Index: 13,05908
Order Now
Specifikation testresultat
Appearance Yellow to orange Powder
Titration Complexometric ≥99.0 %
Residue after ignition ≤0.05 % (substances not reducible with HCOOH)
Iron (Fe) ≤50 ppm