Kadmiumjodid ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Cadmium iodide
A18079.14 A18079.22
A18079.14EA 825 SEK
A18079.14 A18079.22
Kadmiumjodid ≥98%
Kadmiumjodid
Purity: 98% CAS No.: 7790-80-9 Molecular Formula: CdI2 Formula Weight: 366.21 Odour: Odourless Melting Point: 387-389° Boiling Point: 787° Merck Reference: 14,1621 UN No.: 2570 MDL No.: MFCD00010920
Formel: CdI₂
MW: 366.22 g/mol
Kokpunkt: 787 °C
Smältpunkt: 387…389 °C
Densitet: 5.67
Storage Temperature: Omgivande
MDL Number: MFCD00010920
CAS nummer: 7790-80-9
EINECS: 232-223-6
UN: 2570
Merck Index: 12,01657