Zinkklorid 99.99%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
318170100 318170500
ACRO318170100EA 1025 SEK
ACRO318170100 ACRO318170500
Zinkklorid 99.99%, extra pure
Zinkklorid
Formel: ZnCl₂
MW: 136,3 g/mol
Kokpunkt: 732 °C (760 mmHg)
Smältpunkt: 293 °C
Densitet: 2,91 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011295
CAS nummer: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.01 %