Kvicksilverdiklorid 99.5%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Mercuric chloride, Mercury Chloride, Mercury dichloride
201430250 201431000 201435000 201430010
ACRO201430250EA 475 SEK
ACRO201430250 ACRO201431000 ACRO201435000 ACRO201430010
Kvicksilverdiklorid 99.5%
Kvicksilverdiklorid
Formel: HgCl₂
MW: 271.5 g/mol
Kokpunkt: 302 °C (760 mmHg)
Smältpunkt: 277 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011041
CAS nummer: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1 II
Merck Index: 13,05901
Order Now
Specifikation testresultat
Appearance White Crystalline powder or crystals (May contain black particles)
Titration Complexometric ≥99.4 %