Zinkbromid 99.999%, extra pure
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Zinc (II) bromide
212770025 212770100 212770500
ACRO212770025EA 685 SEK
ACRO212770025 ACRO212770100 ACRO212770500
Zinkbromid 99.999%, extra pure
Zinkbromid
Formel: Br₂Zn
MW: 225.2 g/mol
Kokpunkt: 670…680 °C (1013 hPa)
Smältpunkt: 394
Densitet: 4.22 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011294
CAS nummer: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8 II
Merck Index: 13,10182
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Total trace metal impurities ≤0.001 %