Silversulfat, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverantör: Avantor
Fara
Synonyms: Disilver sulphate
1536.0025EA 0 SEK
1536.0025 1536.0250
Silversulfat, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Silversulfat
Formel: Ag₂SO₄
MW: 311.8 g/mol
Kokpunkt: 1085
Smältpunkt: 655
Densitet: 5.45 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003407
CAS nummer: 10294-26-5
EINECS: 233-653-7
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay min. 99.5 %
Iron (Fe) max. 0.001 %
Insoluble Matter and Chloride max. 0.02 %
Nitrate (NO3) max. 0.01 %
Substances not Precipitated by HCl max. 0.03 %