Silversulfat ≥99.999%, Premion®
Leverantör: Thermo Scientific
Fara
Synonyms: Disilver sulphate
12112.06 12112.14 12112.22
12112.06EA 1050 SEK
12112.06 12112.14 12112.22
Silversulfat ≥99.999%, Premion®
Silver sulphate
Purity: 99.999% CAS No.: 10294-26-5 Molecular Formula: Ag2O4S Formula Weight: 311.80 Physical Form: Powder Density: 5.45 Appearance: White Odour: Odourless Melting Point: 657° Boiling Point: 1085° dec. Merck Reference: 14,8529 UN No.: 3077 MDL No.: MFCD00003407
Formel: Ag₂SO₄
MW: 311,8 g/mol
Kokpunkt: 1085 °C (dec.)
Smältpunkt: 657 °C
Densitet: 5,45
Storage Temperature: Omgivande
MDL Number: MFCD00003407
CAS nummer: 10294-26-5
EINECS: 233-653-7
UN: 3077
ADR: 9,III
Merck Index: 14,8529
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=9367d53ccabf2b3585d2283f37dc5dfca622d275&prid=web&clid=SAPDAM