Koppar(II)sulfat, vattenfri, Reagent Grade
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Cupric sulphate
33308.22 33308.36 33308.A3
33308.22EA 1225 SEK
33308.22 33308.36 33308.A3
Koppar(II)sulfat, vattenfri, Reagent Grade
Koppar(II)sulfat
CAS No.: 7758-98-7 Molecular Formula: CuO4S Formula Weight: 159.60 Physical Form: Powder Density: 3.6 Sensitivity: Hygroscopic Appearance: Pale grey Odour: Odourless Melting Point: 560° dec. Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,2653 UN No.: 3288 MDL No.: MFCD00010981
Formel: CuSO₄
MW: 159.61 g/mol
Smältpunkt: >560 °C (dec.)
Densitet: 3.6
Storage Temperature: Omgivande
MDL Number: MFCD00010981
CAS nummer: 7758-98-7
EINECS: 231-847-6
UN: 3288
ADR: 6.1 III
Merck Index: 14,2653