Koppar(I)jodid 99.995%
Leverantör: Thermo Fisher Scientific
Fara
201500050 201500250 201501000
ACRO201500050EA 1375 SEK
ACRO201500050 ACRO201500250 ACRO201501000
Koppar(I)jodid 99.995%
Koppar(I)jodid
Formel: CuI
Kokpunkt: 1290 °C (1013 hPa)
Smältpunkt: 605 °C
Densitet: 5,62 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010978
CAS nummer: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692
Order Now
Specifikation testresultat
Appearance White to grey to light brown Powder
Titration Complexometric 31.0 to 34.0 % (Cu)
Total trace metal impurities ≤50 ppm