Silvercyanid ≥99%
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Argentous cyanide
11424.22
11424.14
11424.22EA
10675
SEK
11424.22
11424.14
Silvercyanid ≥99%
Silvercyanid
Purity: 99%
CAS No.: 506-64-9
Molecular Formula: CAgN
Formula Weight: 133.84
Physical Form: Powder
Density: 3.95
Refractive Index: 1.685
Sensitivity: Light Sensitive
Appearance: White to off-white
Odour: Odourless
Melting Point: 320° dec.
Solubility: Insoluble in water, dilute acids, and alcohol. Soluble in alkali cyanides and boiling concentrated HNO3
Merck Reference: 14,8512
UN No.: 1684
MDL No.: MFCD00003409
Silver plating
Formel:
AgCN MW: 133.89 g/mol Smältpunkt: 320 °C (dec.) Densitet: 3.95 Storage Temperature: Omgivande |
MDL Number:
MFCD00003409 CAS nummer: 506-64-9 EINECS: 208-048-6 UN: 1684 Merck Index: 12,08655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...