Silver perchlorate

Leverantör: GFS CHEMICALS

86.100 86.25
GFSC86.100EA 0 SEK
GFSC86.100 GFSC86.25
Silver perchlorate
Silver perchlorate
Formel: AgClO₄
Smältpunkt: 486 °C
Densitet: 1,688 g/cm³ (25 °C)
MDL Number: MFCD00003400
CAS nummer: 7783-93-9
EINECS: 232-035-4
UN: 1481
ADR: 5.1,II
Merck Index: 12,08667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR