Koppar(I)klorid 97+%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Cuprous chloride, Copper chloride
270520010 270525000 270520025
ACRO270520010EA 2150 SEK
ACRO270520010 ACRO270525000 ACRO270520025
Koppar(I)klorid 97+%, extra pure
Copper (I) chloride
Formel: ClCu
MW: 99 g/mol
Kokpunkt: 1490 °C (760 mmHg)
Smältpunkt: 430 °C
Densitet: 4,14 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00010971
CAS nummer: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8,III
Merck Index: 13,02690
Order Now
Specifikation testresultat
Appearance Light green to grey Crystalline powder
Titration Complexometric 97.0 to 103.0 %
Sulfate (SO4) ≤500 ppm
Arsenic (As) ≤5 ppm
Iron (Fe) ≤100 ppm
Insoluble matter ≤0.05 % (in acid)
Subst nt pptd by H2S(SO4) ≤0.5 %