Koppar(I)klorid 99%, extra pure, renad
Leverantör: Thermo Scientific
Fara
Synonyms: Cuprous chloride, Copper chloride
208390250 208391000 208395000
ACRO208390250EA 1100 SEK
ACRO208390250 ACRO208391000 ACRO208395000
Koppar(I)klorid 99%, extra pure, renad
Copper (I) chloride
Formel: ClCu
MW: 99 g/mol
Kokpunkt: 1490 °C (760 mmHg)
Smältpunkt: 430 °C
Densitet: 4,14 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00010971
CAS nummer: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8,III
Merck Index: 13,02690
Order Now
Specifikation testresultat
Appearance Light green to grey Crystalline powder
Titration Complexometric 98.5 to 101.5 %
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤200 ppm