Koppar(II)bromid, vattenfri 99+%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Cupric bromide, Cuprous bromide
206330050 206331000 206330025 206335000
ACRO206330050EA 530 SEK
ACRO206330050 ACRO206331000 ACRO206330025 ACRO206335000
Koppar(II)bromid, vattenfri 99+%, extra pure
Koppar(II)bromid
Formel: Br₂Cu
MW: 223,35 g/mol
Kokpunkt: 900 °C (760 mmHg)
Smältpunkt: 498 °C
Densitet: 4,71 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010970
CAS nummer: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III
Merck Index: 13,02656
Order Now
Specifikation testresultat
Appearance Dark grey to grey-black Crystalline powder
Titration Iodometric ≥99.0 %
Chloride (Cl) ≤5000 ppm
Sulfate (SO4) ≤2000 ppm
Insoluble matter ≤0.05 % (in water)