1,7-Dihydroxynaphthalene ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Naphthalene-1,7-diol
H28261.14 H28261.06
H28261.14EA 4100 SEK
H28261.14 H28261.06
1,7-Dihydroxynaphthalene ≥97%
1,7-Dihydroxynaphthalene
Purity: 97% CAS No.: 575-38-2 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 180-184° MDL No.: MFCD00035720
Formel: C₁₀H₈O₂
MW: 160,17 g/mol
Kokpunkt: 375 °C (1013 hPa)
Smältpunkt: 180…184 °C
Storage Temperature: Omgivande
MDL Number: MFCD00035720
CAS nummer: 575-38-2
EINECS: 209-383-0