2-Naphthoic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: β-Naphthoic acid
177320250 177321000
ACRO177320250EA 920 SEK
ACRO177320250 ACRO177321000
2-Naphthoic acid 99%
2-Naphthoic acid
Formel: C₁₁H₈O₂
MW: 172,18 g/mol
Kokpunkt: >300 °C (760 mmHg)
Smältpunkt: 183…187 °C
Densitet: 1,382 g/cm³ (20 °C)
Flampunkt: 205 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004101
CAS nummer: 93-09-4
EINECS: 202-217-8
Merck Index: 12,06469
Order Now
Specifikation testresultat
Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 183 to 187 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %