Ammoniumcerium (IV) nitrat ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate
A12882.0B A12882.18 A12882.30 A12882.PG
A12882.0BEA 8625 SEK
A12882.0B A12882.18 A12882.30 A12882.PG
Ammoniumcerium (IV) nitrat ≥98%
Ammoniumcerium (IV) nitrat
Purity: 98+% CAS No.: 16774-21-3 Molecular Formula: H8CeN8O18 Formula Weight: 548.23 Odour: Pungent Merck Reference: 14,1992 MDL No.: MFCD00151121
Formel: (NH₄)₂Ce(NO₃)₆
MW: 548,22 g/mol
Smältpunkt: 107…108 °C
Densitet: 2,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00151121
CAS nummer: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1,II
Merck Index: 14,1992