Ytterbium(III)oxid ≥99.998% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Synonyms: Diytterbium trioxide, Ytterbium oxide
10919.06 10919.14
10919.06EA 2025 SEK
10919.06 10919.14
Ytterbium(III)oxid ≥99.998% (REO, rare earth oxide basis), REacton®
Ytterbium(III) oxide
Purity: 99.998% CAS No.: 1314-37-0 Molecular Formula: Yb2O3 Formula Weight: 394.08 Physical Form: Powder Density: 9.2 Melting Point: 2227° Merck Reference: 14,10105 MDL No.: MFCD00011290
Formel: O₃Yb₂
MW: 394.08 g/mol
Smältpunkt: 2227 °C
Densitet: 9.2
Storage Temperature: Omgivande
MDL Number: MFCD00011290
CAS nummer: 1314-37-0
EINECS: 215-234-0
Merck Index: 14,10105