Terbium(III,IV) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Synonyms: Tetraterbium heptaoxide, Terbium oxide
11207.09 11207.18 11207.A1 11207.30
11207.09EA 6575 SEK
11207.09 11207.18 11207.A1 11207.30
Terbium(III,IV) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium(III,IV) oxide
Purity: 99.9% CAS No.: 12037-01-3 Molecular Formula: Tb4O7 Formula Weight: 747.69 Physical Form: Powder Odour: Odourless Merck Reference: 14,9157 MDL No.: MFCD00083151
Formel: O₇Tb₄
MW: 747,7 g/mol
Smältpunkt: 2340 °C
Storage Temperature: Omgivande
MDL Number: MFCD00083151
CAS nummer: 12037-01-3
EINECS: 234-856-3
Merck Index: 14,9157