Terbium(III,IV) oxide ≥99.998% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Synonyms: Tetraterbium heptaoxide, Terbium oxide
10917.03 10917.06 10917.14 10917.22
10917.03EA 1950 SEK
10917.03 10917.06 10917.14 10917.22
Terbium(III,IV) oxide ≥99.998% (REO, rare earth oxide basis), REacton®
Terbium(III,IV) oxide
Purity: 99.998% CAS No.: 12037-01-3 Molecular Formula: Tb4O7 Formula Weight: 747.69 Physical Form: Powder Odour: Odourless Merck Reference: 14,9157 MDL No.: MFCD00083151
Formel: O₇Tb₄
MW: 747,7 g/mol
Smältpunkt: 2340 °C
Storage Temperature: Omgivande
MDL Number: MFCD00083151
CAS nummer: 12037-01-3
EINECS: 234-856-3
Merck Index: 14,9157