Neodym(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
11248.18 11248.30 11248.A1
11248.18EA 2825 SEK
11248.18 11248.30 11248.A1
Neodym(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium(III) oxide
Purity: 99.9% CAS No.: 1313-97-9 Molecular Formula: Nd2O3 Formula Weight: 336.48 Physical Form: Powder Density: 7.24 Melting Point: 2272° Merck Reference: 14,6450 MDL No.: MFCD00011134
Formel: Nd₂O₃
MW: 336,48 g/mol
Kokpunkt: 3760 °C (1013 hPa)
Smältpunkt: 2272 °C
Densitet: 7,24
Storage Temperature: Omgivande
MDL Number: MFCD00011134
CAS nummer: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,6450