Holmium(III)oxid ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Synonyms: Diholmium trioxide, Holmium oxide
11281.09 11281.18
11281.09EA 3825 SEK
11281.09 11281.18
Holmium(III)oxid ≥99.99% (REO, rare earth oxide basis), REacton®
Holmium(III) oxide
Purity: 99.99% CAS No.: 12055-62-8 Molecular Formula: Ho2O3 Formula Weight: 377.86 Physical Form: Powder Odour: Odourless Merck Reference: 14,4727 MDL No.: MFCD00011053
Formel: Ho₂O₃
MW: 377,86 g/mol
Kokpunkt: 2600 °C (1013 hPa)
Smältpunkt: 2360 °C
Densitet: 3,79 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011053
CAS nummer: 12055-62-8
EINECS: 235-015-3
Merck Index: 14,4727