Holmium(III)oxid ≥99.995% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Synonyms:
Diholmium trioxide, Holmium oxide
10910.03
10910.14
10910.03EA
1450
SEK
10910.03
10910.14
Holmium(III)oxid ≥99.995% (REO, rare earth oxide basis), REacton®
Holmium(III) oxide
Purity: 99.995%
CAS No.: 12055-62-8
Molecular Formula: Ho2O3
Formula Weight: 377.86
Physical Form: Powder
Appearance: Pale brown
Odour: Odourless
Melting Point: 2360°
Merck Reference: 14,4727
MDL No.: MFCD00011053
Formel:
Ho₂O₃ MW: 377,86 g/mol Kokpunkt: 2600 °C (1013 hPa) Smältpunkt: 2360 °C Densitet: 3,79 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00011053 CAS nummer: 12055-62-8 EINECS: 235-015-3 Merck Index: 14,4727 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...