Digadoliniumtrioxid ≥99.9%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Gadolinium (III) oxide
36355.14 36355.22
36355.14EA 3175 SEK
36355.14 36355.22
Digadoliniumtrioxid ≥99.9%
Digadoliniumtrioxid
Purity: 99.9% CAS No.: 12064-62-9 Molecular Formula: Gd2O3 Formula Weight: 362.50 Physical Form: 3-12mm Sintered Lump Density: 7.407 Melting Point: 2330° Merck Reference: 14,4326 MDL No.: MFCD00011026
Formel: Gd₂O₃
MW: 362.5 g/mol
Smältpunkt: 2330 °C
Densitet: 7.407
Storage Temperature: Omgivande
MDL Number: MFCD00011026
CAS nummer: 12064-62-9
EINECS: 235-060-9