Europium(III)oxid ≥99.9% (REO, rare earth oxide basis), nanopowder
Leverantör: Thermo Scientific
44667.06 44667.14
44667.06EA 3450 SEK
44667.06 44667.14
Europium(III)oxid ≥99.9% (REO, rare earth oxide basis), nanopowder
Europium(III) oxide
Purity: 99.9% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: 45-60nm APS Powder, S. A. 14-18m2/g Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997
Formel: Eu₂O₃
MW: 351,92 g/mol
Smältpunkt: 2291 °C
Densitet: 7,42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010997
CAS nummer: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904