Europium(III)oxid ≥99.999% (REO, rare earth oxide basis), nanopowder, REacton®
Leverantör: Thermo Fisher Scientific
44052.14 44052.22
44052.14EA 15550 SEK
44052.14 44052.22
Europium(III)oxid ≥99.999% (REO, rare earth oxide basis), nanopowder, REacton®
Europium(III) oxide
Purity: 99.999% CAS No.: 1308-96-9 Molecular Formula: Eu2O3 Formula Weight: 351.92 Physical Form: 30-50nm APS Powder Sensitivity: Hygroscopic Merck Reference: 14,3904 MDL No.: MFCD00010997
Formel: Eu₂O₃
MW: 351.92 g/mol
Smältpunkt: 2291 °C
Densitet: 7.42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010997
CAS nummer: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,3904