Erbium(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Varning
Synonyms: Dierbium trioxide, Erbium oxide
11310.14 11310.22 11310.A1
11310.14EA 1275 SEK
11310.14 11310.22 11310.A1
Erbium(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium(III) oxide
Purity: 99.9% CAS No.: 12061-16-4 Molecular Formula: Er2O3 Formula Weight: 382.56 Physical Form: Powder Density: 8.64 Melting Point: 2400° Merck Reference: 14,3646 MDL No.: MFCD00010991
Formel: Er₂O₃
MW: 382,52 g/mol
Smältpunkt: 2400 °C
Densitet: 8,64
Storage Temperature: Omgivande
MDL Number: MFCD00010991
CAS nummer: 12061-16-4
EINECS: 235-045-7
Merck Index: 14,3646