Dysprosium(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Synonyms: Didysprosium trioxide, Dysprosium oxide
11319.09 11319.18 11319.30
11319.09EA 1325 SEK
11319.09 11319.18 11319.30
Dysprosium(III)oxid ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium(III) oxide
Purity: 99.9% CAS No.: 1308-87-8 Molecular Formula: Dy2O3 Formula Weight: 373.00 Physical Form: Powder Density: 7.81 Melting Point: 2340° Merck Reference: 14,3482 MDL No.: MFCD00010986
In cermets used as nuclear reaction control rods
Formel: Dy₂O₃
MW: 373 g/mol
Smältpunkt: 2340 °C
Densitet: 7,81
Storage Temperature: Omgivande
MDL Number: MFCD00010986
CAS nummer: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,3482