Ytterbium(III) fluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ytterbium trifluoride, Ytterbium fluoride
11189.03 11189.09 11189.18
11189.03EA 1125 SEK
11189.03 11189.09 11189.18
Ytterbium(III) fluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium(III) fluoride
Purity: 99.99% CAS No.: 13760-80-0 Molecular Formula: YbF3 Formula Weight: 230.04 Physical Form: Lump/Powder Density: 8.2 Melting Point: 1157° UN No.: 3288 MDL No.: MFCD00049615
Formel: F₃Yb
MW: 230.04 g/mol
Kokpunkt: 2227
Smältpunkt: 1157 °C
Densitet: 8.2
Storage Temperature: Omgivande
MDL Number: MFCD00049615
CAS nummer: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1 III