Erbium trifluoride, vattenfri ≥99.9% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Varning
13653.09 13653.18
13653.09EA 2475 SEK
13653.09 13653.18
Erbium trifluoride, vattenfri ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium trifluoride
Purity: 99.9% CAS No.: 13760-83-3 Molecular Formula: ErF3 Formula Weight: 224.28 Physical Form: Powder Density: 6.4 Melting Point: 1350° MDL No.: MFCD00016074
Formel: ErF₃
MW: 224.26 g/mol
Kokpunkt: 2200
Smältpunkt: 1350 °C
Densitet: 6.4
Storage Temperature: Omgivande
MDL Number: MFCD00016074
CAS nummer: 13760-83-3
EINECS: 237-356-3
UN: 3288
ADR: 6.1 III