Erbium trifluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Scientific
Varning
11312.09 11312.18
11312.09EA 3925 SEK
11312.09 11312.18
Erbium trifluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium trifluoride
Purity: 99.99% CAS No.: 13760-83-3 Molecular Formula: ErF3 Formula Weight: 224.28 Physical Form: Granular/Powder Density: 6.4 Melting Point: 1350° MDL No.: MFCD00016074
Formel: ErF₃
MW: 224,26 g/mol
Kokpunkt: 2200 °C (1013 hPa)
Smältpunkt: 1350 °C
Densitet: 6,4
Storage Temperature: Omgivande
MDL Number: MFCD00016074
CAS nummer: 13760-83-3
EINECS: 237-356-3
UN: 3288
ADR: 6.1,III