Neodymium trichloride, vattenfri ≥99.9% (REO, rare earth oxide basis)
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Neodymium (III) chloride
18680.06 18680.14 18680.22
18680.06EA 2050 SEK
18680.06 18680.14 18680.22
Neodymium trichloride, vattenfri ≥99.9% (REO, rare earth oxide basis)
Neodymium trichloride
Purity: 99.9% CAS No.: 10024-93-8 Molecular Formula: NdCl3 Formula Weight: 250.60 Physical Form: -20 Mesh Powder Density: 4.134 Melting Point: 784° Merck Reference: 14,6450 MDL No.: MFCD00011132
Formel: Cl₃Nd
MW: 250.6 g/mol
Kokpunkt: 1600
Smältpunkt: 784 °C
Densitet: 4.134
Storage Temperature: Omgivande
MDL Number: MFCD00011132
CAS nummer: 10024-93-8
EINECS: 233-031-5
Merck Index: 14,6450