Gadolinium trifluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Gadolinium (III) fluoride, Gadolinium(III) bromide
11293.09 11293.18
11293.09EA 1750 SEK
11293.09 11293.18
Gadolinium trifluoride, vattenfri ≥99.99% (REO, rare earth oxide basis), REacton®
Gadolinium trifluoride
Purity: 99.99% CAS No.: 13765-26-9 Molecular Formula: GdF3 Formula Weight: 214.25 Physical Form: Crystalline Density: 7.1 Appearance: White Odour: Odourless Melting Point: 1231° MDL No.: MFCD00016113
Formel: F₃Gd
MW: 214.25 g/mol
Smältpunkt: 1231
Densitet: 7.1
Storage Temperature: Omgivande
MDL Number: MFCD00016113
CAS nummer: 13765-26-9
EINECS: 237-369-4
UN: 3288
ADR: 6.1 III