5-Aminoindole 97%
Leverantör: Thermo Scientific
Varning
Synonyms: Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine
103900010 103900050
ACRO103900010EA 1200 SEK
ACRO103900010 ACRO103900050
5-Aminoindole 97%
5-Aminoindole
Formel: C₈H₈N₂
MW: 132,16 g/mol
Kokpunkt: 190 °C (6 mmHg)
Smältpunkt: 130…134 °C
Storage Temperature: Frys
MDL Number: MFCD00005679
CAS nummer: 5192-03-0
EINECS: 225-977-2
Order Now
Specifikation testresultat
Appearance Off-white to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %