5-Fluoroindole ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5-Fluoro-1H-indole
A15346.03 A15346.06 A15346.14
A15346.03EA 1350 SEK
A15346.03 A15346.06 A15346.14
5-Fluoroindole ≥99%
5-Fluoroindole
Purity: 99% CAS No.: 399-52-0 Molecular Formula: C8H6FN Formula Weight: 135.14 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 42-47° Beilstein Reference: 112350 MDL No.: MFCD00005671
Formel: C₈H₆FN
Kokpunkt: 85 °C (0,5 mmHg)
Smältpunkt: 42…47 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00005671
CAS nummer: 399-52-0
EINECS: 206-917-4