5-Bromo-1-indanone 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Bromo-2,3-dihydro-1H-inden-1-one
309570010 309570050
ACRO309570010EA 700 SEK
ACRO309570010 ACRO309570050
5-Bromo-1-indanone 97%
5-Bromo-1-indanone
Formel: C₉H₇BrO
MW: 211,06 g/mol
Kokpunkt: 280 °C (1013 hPa)
Smältpunkt: 126…130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082718
CAS nummer: 34598-49-7
Order Now
Specifikation testresultat
Appearance White to beige to brown Powder
Infrared spectrum Conforms
Melting point 125 to 130 °C
GC ≥96.0 %