1,1'-Thiocarbonyldiimidazole 90% tech.
Leverantör: Thermo Scientific
Varning
Synonyms: 1,1'-Thiocarbonylbis(imidazole), 1,1'-Thiocarbonylbis-1H-imidazole
168480100 168480250
ACRO168480100EA 1675 SEK
ACRO168480100 ACRO168480250
1,1'-Thiocarbonyldiimidazole 90% tech.
1,1'-Thiocarbonyldiimidazole
Formel: C₇H₆N₄S
MW: 178,22 g/mol
Smältpunkt: 96…103 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005289
CAS nummer: 6160-65-2
EINECS: 228-183-4
Order Now
Specifikation testresultat
Appearance Bright yellow Powder
Infrared spectrum Conforms
Titration with HClO4 ≥88.0 %