1-Metylimidazol ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
A12575.22 A12575.36 A12575.A3
A12575.22EA 920 SEK
A12575.22 A12575.36 A12575.A3
1-Metylimidazol ≥99%
1-Methylimidazole
Purity: 99% CAS No.: 616-47-7 Molecular Formula: C4H6N2 Formula Weight: 82.11 Flash Point: 92°(197°F) Refractive Index: 1.4970 Melting Point: -6° Boiling Point: 195-197° Beilstein Reference: 105197 UN No.: 3267 MDL No.: MFCD00005292
Formel: C₄H₆N₂
MW: 82,11 g/mol
Kokpunkt: 195…197 °C
Smältpunkt: –6 °C
Densitet: 1,031
Flampunkt: 92 °C (197 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005292
CAS nummer: 616-47-7
EINECS: 210-484-7
UN: 3267
ADR: 8,III