2,2'-Dipyridyl disulphide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A11118.06 A11118.14 A11118.22
A11118.06EA 1725 SEK
A11118.06 A11118.14 A11118.22
2,2'-Dipyridyl disulphide ≥98%
2,2'-Dipyridyl disulphide
Purity: 98% CAS No.: 2127-03-9 Molecular Formula: C10H8N2S2 Formula Weight: 220.32 Storage Temperature: 2°C to 8°C Flash Point: 210°(410°F) Melting Point: 56-59° Beilstein Reference: 154629 MDL No.: MFCD00006287
Formel: C₁₀H₈N₂S₂
Kokpunkt: 348 °C (1013 hPa)
Smältpunkt: 56…59 °C
Flampunkt: 210 °C (410 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00006287
CAS nummer: 2127-03-9
EINECS: 218-343-1