Diphenyl disulphide 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2-Diphenyldisulphane, Phenyl disulphide
173710500 173712500
ACRO173710500EA 745 SEK
ACRO173710500 ACRO173712500
Diphenyl disulphide 99%
Diphenyl disulphide
Formel: C₁₂H₁₀S₂
MW: 218,34 g/mol
Kokpunkt: 191…192 °C (15 mmHg)
Smältpunkt: 58…61 °C
Densitet: 1,155 g/cm³ (64.9 °C)
MDL Number: MFCD00003065
CAS nummer: 882-33-7
EINECS: 212-926-4
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance White to light yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 58 to 62 °C
HPLC ≥98.5 %