Dimethyl trisulphide, Supelco®

Leverantör: Merck
Varning

Synonyms: Dimethyltrisulphide

M-Clarity™ quality level = MQ100

79592-10MG-F
79592-10MG-FEA 794 SEK
79592-10MG-F
Dimethyl trisulphide, Supelco®
Dimethyl trisulphide
Formel: C₂H₆S₃
MW: 126.27 g/mol
MDL Number: MFCD00039808
CAS nummer: 3658-80-8
EINECS: 222-910-9
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55069bf33a29b9676ccf91cd34433363dc5fab30&vid=32e3fbd5ab2474d3c85949ca344d7055fdb36e07&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR