Dimetyldisulfid 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Dimethyl disulphide, Methyl disulphide
165590010 165592500 165590025 165590500
ACRO165590010EA 1475 SEK
ACRO165590010 ACRO165592500 ACRO165590025 ACRO165590500
Dimetyldisulfid 99%
Dimetyldisulfid
Formel: C₂H₆S₂
MW: 94,2 g/mol
Kokpunkt: 108…110 °C (760 mmHg)
Smältpunkt: –85 °C
Densitet: 1,0625 g/cm³
Flampunkt: 16 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008561
CAS nummer: 624-92-0
EINECS: 210-871-0
UN: 2381
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5243 to 1.5263 (20°C, 589 nm)
Water ≤0.06 %
GC ≤1 % (Methylmercaptan content)