Dimetylsulfoxid 99.8+% for peptide synthesis
Leverantör: Thermo Scientific
Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
354840025 354845000
ACRO354840025EA 5150 SEK
ACRO354840025 ACRO354845000
Dimetylsulfoxid 99.8+% for peptide synthesis
Dimetylsulfoxid
Formel: (CH₃)₂SO
MW: 78,14 g/mol
Kokpunkt: 189 °C (760 mmHg)
Smältpunkt: 18,4 °C
Densitet: 1,1 g/cm³
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=70006c891d68445bb4e4da63ffc6784c69639512&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Water ≤0.005 % (K.F.)
GC ≥99.80 %
Residue after evaporation ≤0.001 %
Free amine ≤0.0002 %
Acidity (CH3COOH) ≤0.003 %