Tiram ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetrametyltiuramdisulfid
B20063.0E B20063.22 B20063.36
B20063.0EEA 6050 SEK
B20063.0E B20063.22 B20063.36
Tiram ≥97%
Tiram
Purity: 97% CAS No.: 137-26-8 Molecular Formula: C6H12N2S4 Formula Weight: 240.43 Flash Point: 89°(192°F) Melting Point: 152-155° Boiling Point: 129°/20mm Beilstein Reference: 1725821 Merck Reference: 14,9371 UN No.: 3077 MDL No.: MFCD00008325
Formel: C₆H₁₂N₂S₄
Kokpunkt: 129 °C (20 mmHg)
Smältpunkt: 152…155 °C
Densitet: 1,300
Flampunkt: 89 °C (192 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008325
CAS nummer: 137-26-8
EINECS: 205-286-2
UN: 3077
ADR: 9,III
Merck Index: 14,9371