Diallyl disulphide tech. 80% remainder mainly diallyl sulfide and diallyl trisulfide, Technical Grade
Leverantör: Thermo Scientific
Varning
Synonyms: Diallyldisulphide, Allyl disulphide
L11106.06 L11106.14 L11106.22
L11106.06EA 560 SEK
L11106.06 L11106.14 L11106.22
Diallyl disulphide tech. 80% remainder mainly diallyl sulfide and diallyl trisulfide, Technical Grade
Diallyl disulphide
Purity: 80% CAS No.: 2179-57-9 Molecular Formula: C6H10S2 Formula Weight: 146.27 Storage Temperature: 2°C to 8°C Flash Point: 51°(123°F) Refractive Index: 1.5410 Odour: Garlic-like Boiling Point: 180-190° Beilstein Reference: 1699241 UN No.: 1993 MDL No.: MFCD00008656
Formel: C₆H₁₀S₂
MW: 146,28 g/mol
Kokpunkt: 180…190 °C
Densitet: 1,008
Flampunkt: 51 °C (123 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00008656
CAS nummer: 2179-57-9
EINECS: 218-548-6
UN: 1993
ADR: 3,III